Imprimer

Anti-GGPS1 Mouse Monoclonal Antibody [clone: OTI2E7]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA501729EA 603 EUR
ORIGTA501729
Anti-GGPS1 Mouse Monoclonal Antibody [clone: OTI2E7]
Anticorps
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: GGPS1 (geranylgeranyl diphosphate synthase 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR