Imprimer
Anti-GGPS1 Mouse Monoclonal Antibody [clone: OTI3F9]
Fournisseur: OriGene
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ORIGTA501731EA 608 EUR
ORIGTA501731
Anti-GGPS1 Mouse Monoclonal Antibody [clone: OTI3F9]
Anticorps
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:150
Type: Primary
Antigen: GGPS1 (geranylgeranyl diphosphate synthase 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human