Imprimer
Anti-GAS7 Mouse Monoclonal Antibody [clone: OTI2F6]
Fournisseur: OriGene
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ORIGTA501756EA 586 EUR
ORIGTA501756
Anti-GAS7 Mouse Monoclonal Antibody [clone: OTI2F6]
Anticorps
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100
Type: Primary
Antigen: GAS7 (growth arrest-specific 7)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Commander maintenant