Imprimer
Anti-PRKD2 Mouse Monoclonal Antibody [clone: OTI3B12]
Fournisseur: OriGene
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ORIGTA501775EA 608 EUR
ORIGTA501775
Anti-PRKD2 Mouse Monoclonal Antibody [clone: OTI3B12]
Anticorps
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100
Type: Primary
Antigen: PRKD2 (protein kinase D2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Commander maintenant