Imprimer
Anti-PUS7 Mouse Monoclonal Antibody [clone: OTI5F12]
Fournisseur: OriGene
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ORIGTA501815EA 603 EUR
ORIGTA501815
Anti-PUS7 Mouse Monoclonal Antibody [clone: OTI5F12]
Anticorps
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100
Type: Primary
Antigen: PUS7 (pseudouridylate synthase 7 (putative))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Commander maintenant