Imprimer

Anti-GSTT2 Mouse Monoclonal Antibody [clone: OTI6B7]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA501830EA 581 EUR
ORIGTA501830
Anti-GSTT2 Mouse Monoclonal Antibody [clone: OTI6B7]
Anticorps
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100; Immunochemistry: 1:50-1:150

Type: Primary
Antigen: GSTT2 (glutathione S-transferase theta 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR