Imprimer

Anti-SULT1A1 Mouse Monoclonal Antibody [clone: OTI5F1]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA501837EA 608 EUR
ORIGTA501837
Anti-SULT1A1 Mouse Monoclonal Antibody [clone: OTI5F1]
Anticorps
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: SULT1A1 (sulfotransferase family, cytosolic, 1A, phenol-preferring, member 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR