Imprimer

Anti-HSPA6 Mouse Monoclonal Antibody [clone: OTI2C2]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA501839EA 593 EUR
ORIGTA501839
Anti-HSPA6 Mouse Monoclonal Antibody [clone: OTI2C2]
Anticorps
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: HSPA6 (heat shock 70kDa protein 6 (HSP70B'))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR