Imprimer

Anti-ALDH1L1 Mouse Monoclonal Antibody [clone: OTI4C11]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA501851EA 603 EUR
ORIGTA501851
Anti-ALDH1L1 Mouse Monoclonal Antibody [clone: OTI4C11]
Anticorps
Recommended Dilutions: Western Blot: 1:500-2000; Flow Cytometry: 1:100

Type: Primary
Antigen: ALDH1L1 (aldehyde dehydrogenase 1 family, member L1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR