Imprimer
Anti-PPWD1 Mouse Monoclonal Antibody [clone: OTI1B2]
Fournisseur: OriGene
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ORIGTA501891EA 581 EUR
ORIGTA501891
Anti-PPWD1 Mouse Monoclonal Antibody [clone: OTI1B2]
Anticorps
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100
Type: Primary
Antigen: PPWD1 (peptidylprolyl isomerase domain and WD repeat containing 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human