Imprimer

Anti-OTUB2 Mouse Monoclonal Antibody [clone: OTI2F6]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA501944EA 603 EUR
ORIGTA501944
Anti-OTUB2 Mouse Monoclonal Antibody [clone: OTI2F6]
Anticorps
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100

Type: Primary
Antigen: OTUB2 (OTU deubiquitinase, ubiquitin aldehyde binding 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR