Imprimer

Anti-SAMHD1 Mouse Monoclonal Antibody [clone: OTI2A5]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA501956EA 603 EUR
ORIGTA501956
Anti-SAMHD1 Mouse Monoclonal Antibody [clone: OTI2A5]
Anticorps
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: SAMHD1 (SAM domain and HD domain 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR