Imprimer
Anti-CNDP1 Mouse Monoclonal Antibody [clone: OTI2D2]
Fournisseur: OriGene
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ORIGTA502093EA 603 EUR
ORIGTA502093
Anti-CNDP1 Mouse Monoclonal Antibody [clone: OTI2D2]
Anticorps
Recommended Dilutions: Western Blot: 1:500
Type: Primary
Antigen: CNDP1 (carnosine dipeptidase 1 (metallopeptidase M20 family))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human