Imprimer

Anti-DLG3 Mouse Monoclonal Antibody [clone: OTI5F1]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA502191EA 603 EUR
ORIGTA502191
Anti-DLG3 Mouse Monoclonal Antibody [clone: OTI5F1]
Anticorps
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: DLG3 (discs, large homolog 3 (Drosophila))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR