Imprimer

Anti-DNAJB1 Mouse Monoclonal Antibody [clone: OTI1E9]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA502194EA 603 EUR
ORIGTA502194
Anti-DNAJB1 Mouse Monoclonal Antibody [clone: OTI1E9]
Anticorps
Recommended Dilutions: Western Blot: 1:1000-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: DNAJB1 (DnaJ (Hsp40) homolog, subfamily B, member 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR