Imprimer

Anti-LMAN1 Mouse Monoclonal Antibody [clone: OTI1C5]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA502237EA 603 EUR
ORIGTA502237
Anti-LMAN1 Mouse Monoclonal Antibody [clone: OTI1C5]
Anticorps
  • Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: LMAN1 (lectin, mannose-binding, 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR