Imprimer

Anti-PACSIN3 Mouse Monoclonal Antibody [clone: OTI1E1]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA502260EA 581 EUR
ORIGTA502260
Anti-PACSIN3 Mouse Monoclonal Antibody [clone: OTI1E1]
Anticorps
Recommended Dilutions: Western Blot: 1:500-2000; Flow Cytometry: 1:100

Type: Primary
Antigen: PACSIN3 (protein kinase C and casein kinase substrate in neurons 3)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR