Imprimer
Anti-BCHE Mouse Monoclonal Antibody [clone: OTI4C12]
Fournisseur: OriGene
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ORIGTA502288EA 603 EUR
ORIGTA502288
Anti-BCHE Mouse Monoclonal Antibody [clone: OTI4C12]
Anticorps
  • Recommended Dilutions: Western Blot: 1:2000
Type: Primary
Antigen: BCHE (butyrylcholinesterase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human