Imprimer

Anti-RDH14 Mouse Monoclonal Antibody [clone: OTI6C3]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA502315EA 603 EUR
ORIGTA502315
Anti-RDH14 Mouse Monoclonal Antibody [clone: OTI6C3]
Anticorps
Recommended Dilutions: Western Blot: 1:200-500

Type: Primary
Antigen: RDH14 (retinol dehydrogenase 14 (all-trans/9-cis/11-cis))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR