Imprimer

Anti-NUDT6 Mouse Monoclonal Antibody [clone: OTI7F4]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA502332EA 603 EUR
ORIGTA502332
Anti-NUDT6 Mouse Monoclonal Antibody [clone: OTI7F4]
Anticorps
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: NUDT6 (nudix (nucleoside diphosphate linked moiety X)-type motif 6)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR