Imprimer

Anti-PDE2A Mouse Monoclonal Antibody [clone: OTI2F1]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA502354EA 603 EUR
ORIGTA502354
Anti-PDE2A Mouse Monoclonal Antibody [clone: OTI2F1]
Anticorps
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: PDE2A (phosphodiesterase 2A, cGMP-stimulated)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR