Imprimer

Anti-MCTS1 Mouse Monoclonal Antibody [clone: OTI2G2]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

TA502418
ORIGTA502418EA 616 EUR
ORIGTA502418
Anti-MCTS1 Mouse Monoclonal Antibody [clone: OTI2G2]
Anticorps
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:150

Type: Primary
Antigen: MCTS1 (malignant T cell amplified sequence 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR