Imprimer

Anti-UBL4A Mouse Monoclonal Antibody [clone: OTI3C12]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA502448EA 603 EUR
ORIGTA502448
Anti-UBL4A Mouse Monoclonal Antibody [clone: OTI3C12]
Anticorps
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: UBL4A (ubiquitin-like 4A)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR