Imprimer

Anti-PNMA3 Mouse Monoclonal Antibody [clone: OTI3F4]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA502457EA 603 EUR
ORIGTA502457
Anti-PNMA3 Mouse Monoclonal Antibody [clone: OTI3F4]
Anticorps
Recommended Dilutions: Western Blot: 1:500

Type: Primary
Antigen: PNMA3 (paraneoplastic Ma antigen 3)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR