Imprimer

Anti-CAT Mouse Monoclonal Antibody [clone: OTI3C3]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA502497EA 603 EUR
ORIGTA502497
Anti-CAT Mouse Monoclonal Antibody [clone: OTI3C3]
Anticorps
  • Recommended Dilutions: Western Blot: 1:200-500; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: CAT (catalase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR