Imprimer
Anti-RASSF1 Mouse Monoclonal Antibody [clone: OTI8A11]
Fournisseur: OriGene
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ORIGTA502512EA 603 EUR
ORIGTA502512
Anti-RASSF1 Mouse Monoclonal Antibody [clone: OTI8A11]
Anticorps
Recommended Dilutions: Western Blot: 1:500; Immunofluorescence: 1:100
Type: Primary
Antigen: RASSF1 (Ras association (RalGDS/AF-6) domain family member 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human