Imprimer
Anti-POLR2E Mouse Monoclonal Antibody [clone: OTI3B5]
Fournisseur: OriGene
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ORIGTA502548EA 608 EUR
ORIGTA502548
Anti-POLR2E Mouse Monoclonal Antibody [clone: OTI3B5]
Anticorps
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Flow Cytometry: 1:100
Type: Primary
Antigen: POLR2E (polymerase (RNA) II (DNA directed) polypeptide E, 25kDa)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Commander maintenant