Imprimer

Anti-NDUFB9 Mouse Monoclonal Antibody [clone: OTI4F8]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA502553EA 603 EUR
ORIGTA502553
Anti-NDUFB9 Mouse Monoclonal Antibody [clone: OTI4F8]
Anticorps
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:500

Type: Primary
Antigen: NDUFB9 (NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 9, 22kDa)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR