Imprimer

Anti-HDHD1 Mouse Monoclonal Antibody [clone: OTI3E5]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA502557EA 603 EUR
ORIGTA502557
Anti-HDHD1 Mouse Monoclonal Antibody [clone: OTI3E5]
Anticorps
  • Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: HDHD1 (haloacid dehalogenase-like hydrolase domain containing 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR