Imprimer
Anti-ANKRD53 Mouse Monoclonal Antibody [clone: OTI1C10]
Fournisseur: OriGene
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ORIGTA502600EA 603 EUR
ORIGTA502600
Anti-ANKRD53 Mouse Monoclonal Antibody [clone: OTI1C10]
Anticorps
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100
Type: Primary
Antigen: ANKRD53 (ankyrin repeat domain 53)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human