Imprimer

Anti-DSTN Mouse Monoclonal Antibody [clone: OTI2F7]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA502608EA 603 EUR
ORIGTA502608
Anti-DSTN Mouse Monoclonal Antibody [clone: OTI2F7]
Anticorps
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: DSTN (destrin (actin depolymerizing factor))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR