Imprimer

Anti-BECN1 Mouse Monoclonal Antibody [clone: OTI2D4]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA502618EA 603 EUR
ORIGTA502618
Anti-BECN1 Mouse Monoclonal Antibody [clone: OTI2D4]
Anticorps
  • Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: BECN1 (beclin 1, autophagy related)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR