Imprimer
Anti-BECN1 Mouse Monoclonal Antibody [clone: OTI2A7]
Fournisseur: OriGene
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ORIGTA502619EA 603 EUR
ORIGTA502619
Anti-BECN1 Mouse Monoclonal Antibody [clone: OTI2A7]
Anticorps
  • Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100
Type: Primary
Antigen: BECN1 (beclin 1, autophagy related)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human