Imprimer
Anti-DSTN Mouse Monoclonal Antibody [clone: OTI1D8]
Fournisseur: OriGene
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ORIGTA502654EA 608 EUR
ORIGTA502654
Anti-DSTN Mouse Monoclonal Antibody [clone: OTI1D8]
Anticorps
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100
Type: Primary
Antigen: DSTN (destrin (actin depolymerizing factor))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Commander maintenant