Imprimer

Anti-SULT1C2 Mouse Monoclonal Antibody [clone: OTI5H1]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA502678EA 603 EUR
ORIGTA502678
Anti-SULT1C2 Mouse Monoclonal Antibody [clone: OTI5H1]
Anticorps
Recommended Dilutions: Western Blot: 1:500-2000

Type: Primary
Antigen: SULT1C2 (sulfotransferase family, cytosolic, 1C, member 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR