Imprimer

Anti-C2orf40 Mouse Monoclonal Antibody [clone: OTI3B10]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

TA502699
ORIGTA502699EA 598 EUR
ORIGTA502699
Anti-C2orf40 Mouse Monoclonal Antibody [clone: OTI3B10]
Anticorps
Recommended Dilutions: Western Blot: 1:500; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: C2orf40 (chromosome 2 open reading frame 40)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR