Imprimer

Anti-NDUFA7 Mouse Monoclonal Antibody [clone: OTI2C4]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA502749EA 603 EUR
ORIGTA502749
Anti-NDUFA7 Mouse Monoclonal Antibody [clone: OTI2C4]
Anticorps
Recommended Dilutions: Western Blot: 1:500-2000

Type: Primary
Antigen: NDUFA7 (NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 7, 14.5kDa)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR