Imprimer

Anti-PLEKHA3 Mouse Monoclonal Antibody [clone: OTI5D11]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA502927EA 586 EUR
ORIGTA502927
Anti-PLEKHA3 Mouse Monoclonal Antibody [clone: OTI5D11]
Anticorps
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: PLEKHA3 (pleckstrin homology domain containing, family A (phosphoinositide binding specific) member 3)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR