Imprimer

Anti-NDUFAF7 Mouse Monoclonal Antibody [clone: OTI6F10]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA502957EA 603 EUR
ORIGTA502957
Anti-NDUFAF7 Mouse Monoclonal Antibody [clone: OTI6F10]
Anticorps
Recommended Dilutions: Western Blot: 1:500; Flow Cytometry: 1:100

Type: Primary
Antigen: NDUFAF7 (NADH dehydrogenase (ubiquinone) complex I, assembly factor 7)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR