Imprimer

Anti-LSM1 Mouse Monoclonal Antibody [clone: OTI1F5]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA503105EA 603 EUR
ORIGTA503105
Anti-LSM1 Mouse Monoclonal Antibody [clone: OTI1F5]
Anticorps
  • Recommended Dilutions: Western Blot: 1:500

Type: Primary
Antigen: LSM1 (LSM1, U6 small nuclear RNA associated)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR