Imprimer

Anti-ALDH3A2 Mouse Monoclonal Antibody [clone: OTI1D1]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA503165EA 603 EUR
ORIGTA503165
Anti-ALDH3A2 Mouse Monoclonal Antibody [clone: OTI1D1]
Anticorps
Recommended Dilutions: Western Blot: 1:500

Type: Primary
Antigen: ALDH3A2 (aldehyde dehydrogenase 3 family, member A2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR