Imprimer
Anti-ACY1 Mouse Monoclonal Antibody [clone: OTI2B5]
Fournisseur: OriGene
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ORIGTA503191EA 603 EUR
ORIGTA503191
Anti-ACY1 Mouse Monoclonal Antibody [clone: OTI2B5]
Anticorps
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100
Type: Primary
Antigen: ACY1 (aminoacylase 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human