Imprimer

Anti-PSMD2 Mouse Monoclonal Antibody [clone: OTI1G6]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA503217EA 608 EUR
ORIGTA503217
Anti-PSMD2 Mouse Monoclonal Antibody [clone: OTI1G6]
Anticorps
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: PSMD2 (proteasome (prosome, macropain) 26S subunit, non-ATPase, 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR