Imprimer

Anti-PNPO Mouse Monoclonal Antibody [clone: OTI3B3]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA503249EA 608 EUR
ORIGTA503249
Anti-PNPO Mouse Monoclonal Antibody [clone: OTI3B3]
Anticorps
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100

Type: Primary
Antigen: PNPO (pyridoxamine 5'-phosphate oxidase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR