Imprimer

Anti-DHFRL1 Mouse Monoclonal Antibody [clone: OTI1E2]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA503259EA 586 EUR
ORIGTA503259
Anti-DHFRL1 Mouse Monoclonal Antibody [clone: OTI1E2]
Anticorps
  • Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100

Type: Primary
Antigen: DHFRL1 (dihydrofolate reductase-like 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR