Imprimer

Anti-NDEL1 Mouse Monoclonal Antibody [clone: OTI5C2]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA503313EA 603 EUR
ORIGTA503313
Anti-NDEL1 Mouse Monoclonal Antibody [clone: OTI5C2]
Anticorps
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: NDEL1 (nudE neurodevelopment protein 1-like 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR