Imprimer
Anti-UBOX5 Mouse Monoclonal Antibody [clone: OTI1C5]
Fournisseur: OriGene
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ORIGTA503340EA 603 EUR
ORIGTA503340
Anti-UBOX5 Mouse Monoclonal Antibody [clone: OTI1C5]
Anticorps
Recommended Dilutions: Western Blot: 1:200-500; Immunochemistry: 1:150; Flow Cytometry: 1:100
Type: Primary
Antigen: UBOX5 (U-box domain containing 5)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human