Imprimer

Anti-GTF2F1 Mouse Monoclonal Antibody [clone: OTI1D5]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA503372EA 616 EUR
ORIGTA503372
Anti-GTF2F1 Mouse Monoclonal Antibody [clone: OTI1D5]
Anticorps
Recommended Dilutions: Western Blot: 1:1000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: GTF2F1 (general transcription factor IIF, polypeptide 1, 74kDa)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR