Imprimer

Anti-PGAM2 Mouse Monoclonal Antibody [clone: OTI4H1]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA503399EA 603 EUR
ORIGTA503399
Anti-PGAM2 Mouse Monoclonal Antibody [clone: OTI4H1]
Anticorps
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: PGAM2 (phosphoglycerate mutase 2 (muscle))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR