Imprimer

Anti-PDE4B Mouse Monoclonal Antibody [clone: OTI2B10]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA503446EA 603 EUR
ORIGTA503446
Anti-PDE4B Mouse Monoclonal Antibody [clone: OTI2B10]
Anticorps
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: PDE4B (phosphodiesterase 4B, cAMP-specific)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR